(±)-2-Methylvaleric acid ≥98% Kosher

Danger

Synonyms: (±)-2-Methylpentanoic acid, 2-Methylpentanoic Acid, 2-Methylvaleric Acid

W275409-1KG-K W275409-5KG-K W275409-100G-K
220232-636EA 264.62 USD
220232-636 220232-637 220232-635
(±)-2-Methylvaleric acid ≥98% Kosher
(±)-2-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00002671
CAS Number: 97-61-0
EINECS: 202-594-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR