Methyl isovalerate ≥99%

Synonyms: Isopropyl acetic acid methyl ester, Isovaleric acid methyl ester, 3-Methylbutyric acid emthyl ester, Methyl isopentanoate, 3-Methylbutanoic acid methyl ester

W275301-1KG-K W275301-4KG-K W275301-9KG-K
220232-631EA 280 USD
220232-631 220232-632 220232-633
Methyl isovalerate ≥99%
Methyl isovalerate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 0.90065 g/cm³ (0 °C)
MDL Number: MFCD00042866
CAS Number: 556-24-1
EINECS: 209-117-3
UN: 2400
ADR: 3,II
Merck Index: 12,06166

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR