Methyl valerate ≥99% Kosher

Synonyms: Valeric acid methyl ester, Methyl Pentanoate

W275204-1KG-K W275204-4KG-K W275204-9KG-K
220232-627EA 198.46 USD
220232-627 220232-628 220232-629
Methyl valerate ≥99% Kosher
Methyl valerate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 126.5 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS Number: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR