4-Methyl-1-phenyl-2-pentanone ≥92%, halal

Synonyms: 4-Methyl-1-phenylpentan-2-one

W274003-1KG-K W274003-100G-K
220232-602EA 807.69 USD
220232-602 220232-601
4-Methyl-1-phenyl-2-pentanone ≥92%, halal
4-Methyl-1-phenyl-2-pentanone
Formula: (C₆H₅)CH₂C(O)CH₂CH(CH₃)₂
MW: 176.26 g/mol
MDL Number: MFCD00015639
CAS Number: 5349-62-2
EINECS: 226-316-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR