(±)-2-Phenyl-1-propanol ≥97%

Synonyms: 2-Phenylpropan-1-ol

W273201-5KG W273201-1KG W273201-10KG
220232-595EA 3969.23 USD
220232-595 220232-594 220232-593
(±)-2-Phenyl-1-propanol ≥97%
(±)-2-Phenyl-1-propanol
Formula: C₆H₅CH(CH₃)CH₂OH
MW: 136.19 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: -37 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004736
CAS Number: 1123-85-9
EINECS: 214-379-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR