Methyl octanoate ≥99%

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate

W272809-20KG-K W272809-1KG-K W272809-9KG-K
220232-582EA 128 USD
220232-582 220232-581 220232-583
Methyl octanoate ≥99%
Methyl octanoate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –36.7 °C
Density: 0.877 g/cm³ (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00009551
CAS Number: 111-11-5
EINECS: 203-835-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR