2-Nonynoic acid methyl ester ≥99%, halal

Synonyms: Methyl non-2-ynoate, Methyl 2-nonynoate

W272604-5KG-K W272604-1KG-K W272604-100G-K
220232-579EA 827.69 USD
220232-579 220232-578 220232-577
2-Nonynoic acid methyl ester ≥99%, halal
2-Nonynoic acid methyl ester
Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
MDL Number: MFCD00009547
CAS Number: 111-80-8
EINECS: 203-909-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR