2-Acetylnaphthalene Kosher

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

W272302-100G-K W272302-1KG-K
220232-565EA 117.69 USD
220232-565 220232-566
2-Acetylnaphthalene Kosher
2-Acetylnaphthalene
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.1171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR