Methyl myristate ≥98%

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate

W272205-1KG-K W272205-20KG-K W272205-8KG-K
220232-561EA 181.54 USD
220232-561 220232-562 220232-563
Methyl myristate ≥98%
Methyl myristate
Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.86715 g/cm³ (20 °C)
Flash Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR