(±)-Methyl-2-methylbutyrate ≥98%, FG

Synonyms: Methyl2-methylbutyrate

W271926-1KG-K W271926-4KG-K W271926-100G-K
220232-483EA 678.46 USD
220232-483 220231-639 220232-482
(±)-Methyl-2-methylbutyrate ≥98%, FG
(±)-Methyl-2-methylbutyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00009335
CAS Number: 868-57-5
EINECS: 212-778-0
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR