Methyl-o-anisate ≥97%

Synonyms: 2-Methoxybenzoic acid methyl ester, Methyl 2-methoxybenzoate, o-Anisic acid methyl ester, 2-Anisic acid methyl ester

W271705-1KG-K W271705-100G-K
220231-794EA 1584.62 USD
220231-794 220231-793
Methyl-o-anisate ≥97%
Methyl-o-anisate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00008423
CAS Number: 606-45-1
EINECS: 210-118-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR