Methyl laurate ≥98%

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

W271500-1KG W271500-4KG W271500-9KG
220231-789EA 156.92 USD
220231-789 220231-790 220231-791
Methyl laurate ≥98%
Methyl laurate
Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 4.5 °C
Density: 0.8694 g/cm³ (20 °C)
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR