(±)-2-Methylbutyric acid ≥98%, halal

Synonyms: (±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid

W269506-5KG-K W269506-25KG-K W269506-1KG-K
220232-433EA 640 USD
220232-433 220232-432 220232-431
(±)-2-Methylbutyric acid ≥98%, halal
(±)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 177 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR