Methyl isobutyrate ≥99%

W269409-1KG-K W269409-9KG-K W269409-20KG-K
220232-426EA 144.62 USD
220232-426 220232-428 220232-427
Methyl isobutyrate ≥99%
Methyl isobutyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00008914
CAS Number: 547-63-7
EINECS: 208-929-5
UN: 1237
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR