Methyl butyrate ≥98%, FG

Synonyms: Butyric acid methyl ester, Butanoic acid methyl ester

W269328-1KG-K W269328-100G-K W269328-4KG-K
220232-423EA 664.62 USD
220232-423 220232-422 220232-424
Methyl butyrate ≥98%, FG
Methyl butyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: -84 °C
Density: 0.8984 g/cm³ (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR