Methyl butyrate ≥98%

Synonyms: Butyric acid methyl ester, Butanoic acid methyl ester

W269301-20KG-K W269301-5KG-K W269301-1KG-K
220232-419EA 830.77 USD
220232-419 220232-420 220232-418
Methyl butyrate ≥98%
Methyl butyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: -84 °C
Density: 0.8984 g/cm³ (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR