4-Methylanisole ≥99% FCC

Warning

Synonyms: p-Methylanisole

W268100-5KG-K W268100-1KG-K
220231-604EA 139.23 USD
220231-604 220231-603
4-Methylanisole ≥99% FCC
4-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00008413
CAS Number: 104-93-8
EINECS: 203-253-7
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR