2-Methylanisole ≥99%, FG

Synonyms: o-Cresol methyll ether, 2-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Methoxytoluene

W268003-1KG-K W268003-5KG-K W268003-100G-K
220231-599EA 723.08 USD
220231-599 220231-600 220231-598
2-Methylanisole ≥99%, FG
2-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 172 °C (1013 hPa)
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR