Methyl acetate Kosher

Danger

Synonyms: Acetic acid methyl ester

W267619-1KG-K W267619-5KG-K
220229-734EA 350.77 USD
220229-734 220229-735
Methyl acetate Kosher
Methyl acetate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56.9 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 13,06038

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR