4-Methoxyphenylacetone ≥97%

Synonyms: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone

W267406-100G-K W267406-1KG-K
220232-383EA 80 USD
220232-383 220232-384
4-Methoxyphenylacetone ≥97%
4-Methoxyphenylacetone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: -15 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008773
CAS Number: 122-84-9
EINECS: 204-578-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR