2-Methoxy-p-cresol ≥97% Kosher

Warning

Synonyms: Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol

W267112-1KG-K W267112-100G-K
220232-376EA 2538.46 USD
220232-376 220232-375
2-Methoxy-p-cresol ≥97% Kosher
2-Methoxy-p-cresol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 219…222 °C (1013 hPa)
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 13,02599

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR