Maltol, halal

Warning

Synonyms: 3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone

W265624-1KG-K W265624-5KG-K W265624-100G-K
220232-318EA 284.62 USD
220232-318 220232-319 220232-317
Maltol, halal
Maltol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 285 °C (1013 hPa)
Density: 1.453 g/cm³ (20 °C)
MDL Number: MFCD00006578
CAS Number: 118-71-8
EINECS: 204-271-8
Merck Index: 13,05735

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR