Lauric acid ≥98% FCC

Warning

Synonyms: Dodecanoic acid, Dodecylic acid

W261408-1KG-K W261408-25KG-K W261408-10KG-K W261408-5KG-K
220232-227EA 117.69 USD
220232-227 220232-228 220232-226 220232-229
Lauric acid ≥98% FCC
Lauric acid
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR