Lactic acid (natural) ≥85% in aqueous solution

Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

W261114-5KG-K W261114-10KG-K W261114-1KG-K
220232-224EA 463.08 USD
220232-224 220232-222 220232-223
Lactic acid (natural) ≥85% in aqueous solution
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Boiling Pt: 122 °C (20 hPa)
Density: 1.11 to 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR