Hexyl hexanoate ≥97%, halal

Synonyms: Hexanoic acid hexyl ester

W257206-20KG-K W257206-1KG-K W257206-9KG-K
220232-178EA 784.62 USD
220232-178 220232-177 220232-179
Hexyl hexanoate ≥97%, halal
Hexyl hexanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245…246 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.863 g/cm³ (25 °C)
MDL Number: MFCD00053808
CAS Number: 6378-65-0
EINECS: 228-952-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR