Heptyl butyrate ≥98% Kosher

W254908-4KG-K W254908-1KG-K W254908-9KG-K
220231-748EA 1040 USD
220231-748 220231-747 220231-749
Heptyl butyrate ≥98% Kosher
Heptyl butyrate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 225…226 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.864 g/cm³ (25 °C)
MDL Number: MFCD00048908
CAS Number: 5870-93-9
EINECS: 227-526-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR