1-Heptanol ≥97% Kosher

Warning

Synonyms: Heptyl alcohol

W254800-20KG-K W254800-1KG-K W254800-4KG-K
220231-743EA 583.08 USD
220231-743 220231-742 220231-744
1-Heptanol ≥97% Kosher
1-Heptanol
Formula: H₃C(CH₂)₅CH₂OH
MW: 116.2 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR