Heptyl acetate ≥98% Kosher

Synonyms: Acetic acid heptyl ester

W254711-100G-K W254711-1KG-K
220231-738EA 243.08 USD
220231-738 220231-739
Heptyl acetate ≥98% Kosher
Heptyl acetate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –50.2 °C
Density: 0.8891 g/cm³ (0 °C)
MDL Number: MFCD00027311
CAS Number: 112-06-1
EINECS: 203-932-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR