Heptyl acetate ≥98%, halal

Synonyms: Acetic acid heptyl ester

W254703-1KG-K W254703-9KG-K W254703-4KG-K
220231-733EA 119.23 USD
220231-733 220231-736 220231-735
Heptyl acetate ≥98%, halal
Heptyl acetate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –50.2 °C
Density: 0.8891 g/cm³ (0 °C)
MDL Number: MFCD00027311
CAS Number: 112-06-1
EINECS: 203-932-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR