Glycerine ≥99%

W252506-1KG-K W252506-10KG-K W252506-25KG-K
220232-033EA 117.69 USD
220232-033 220232-032 220232-034
Glycerine ≥99%
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=d004eeb9f845a013dfcafd0b44d36127512f5d06&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR