Geranyl butyrate FCC

Synonyms: Butyric acid geranyl ester, 3,7-Dimethylocta-2,6-dien-1-yl butyrate, Butanoic acid geranyl ester

W251208-9KG-K W251208-1KG-K
220231-579EA 1435.38 USD
220231-579 220231-577
Geranyl butyrate FCC
Geranyl butyrate
Formula: C₁₄H₂₄O₂
MW: 224.34 g/mol
MDL Number: MFCD00036538
CAS Number: 106-29-6
EINECS: 203-381-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR