Geraniol FCC

Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

W250708-9KG-K W250708-1KG-K W250708-20KG-K
220231-560EA 655.38 USD
220231-560 220231-558 220231-559
Geraniol FCC
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR