Furfuryl alcohol, halal

Danger

Synonyms: Furfurol

W249106-25KG-K W249106-5KG-K W249106-1KG-K
220231-918EA 775.38 USD
220231-918 220231-919 220231-917
Furfuryl alcohol, halal
Furfuryl alcohol
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 13,04326

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR