2-Furaldehyde Kosher

Danger

Synonyms: 2-Furfurol, Furfural

W248924-10KG-K W248924-1KG-K W248924-5KG-K
220231-904EA 878.46 USD
220231-904 220231-905 220231-906
2-Furaldehyde Kosher
2-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: -39 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1,II
Merck Index: 13,04325

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR