Fumaric acid ≥99% FCC

Warning

Synonyms: trans-1,2-Ethylentricarboxylic acid

W248800-1KG-K W248800-25KG-K
220231-896EA 117.69 USD
220231-896 220231-897
Fumaric acid ≥99% FCC
Fumaric acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 287 °C
Density: 1.625 g/cm³ (20 °C)
Flash Pt: 273 °C
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR