Eugenol ≥98% Kosher

Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

W246719-1KG-K W246719-5KG-K W246719-10KG-K
220231-879EA 167.69 USD
220231-879 220231-880 220231-878
Eugenol ≥98% Kosher
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR