Eugenol ≥98%, FG

Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

W246700-5KG-K W246700-1KG-K
220231-876EA 680 USD
220231-876 220231-875
Eugenol ≥98%, FG
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR