Ethyl isovalerate ≥98%, halal

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

W246328-100G-K W246328-1KG-K
220231-854EA 116.92 USD
220231-854 220231-855
Ethyl isovalerate ≥98%, halal
Ethyl isovalerate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 131…133 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00009203
CAS Number: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 13,03849

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR