Ethyl isovalerate ≥98%, FG

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

W246301-1KG-K W246301-20KG-K W246301-4KG-K
220231-850EA 114.46 USD
220231-850 220231-851 220231-852
Ethyl isovalerate ≥98%, FG
Ethyl isovalerate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 131 - 133 °C (1013 hPa)
Melting Pt: -99 °C
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00009203
CAS Number: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 13,03849

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR