Ethyl valerate ≥98%

Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester

W246204-1KG-K W246204-20KG-K W246204-9KG-K
220231-846EA 107.69 USD
220231-846 220231-847 220231-848
Ethyl valerate ≥98%
Ethyl valerate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: -91 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 12,10042

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR