Ethyl sorbate

Synonyms: Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate

W245909-100G-K W245909-1KG-K W245909-5KG-K
220231-834EA 121.38 USD
220231-834 220231-835 220231-836
Ethyl sorbate
Ethyl sorbate
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 195.4 °C (1013 hPa)
Density: 0.9405 g/cm³ (15.3 °C)
MDL Number: MFCD00009296
CAS Number: 2396-84-1
EINECS: 219-258-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR