Ethyl propionate ≥97% Kosher

Danger

Synonyms: Propionic acid ethyl ester

W245615-100G-K W245615-1KG-K W245615-5KG-K
220231-817EA 145.85 USD
220231-817 220231-818 220231-819
Ethyl propionate ≥97% Kosher
Ethyl propionate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3
EINECS: 203-291-4
UN: 1195
ADR: 3,II
Merck Index: 13,03878

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR