Ethyl-3-phenylpropionate ≥98%

Synonyms: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester

W245518-250G W245518-1KG
220231-810EA 373.85 USD
220231-810 220231-809
Ethyl-3-phenylpropionate ≥98%
Ethyl-3-phenylpropionate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 247 °C (1013 hPa)
Density: 1.0343 g/cm³ (0 °C)
MDL Number: MFCD00009206
CAS Number: 2021-28-5
EINECS: 217-966-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR