Ethyl palmitate ≥95%, halal

Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester

W245100-1KG-K W245100-10KG-K W245100-5KG-K W245100-250G-K
220231-722EA 436.92 USD
220231-722 220231-721 220231-724 220231-723
Ethyl palmitate ≥95%, halal
Ethyl palmitate
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 24 °C
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00008996
CAS Number: 628-97-7
EINECS: 211-064-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR