Ethyl pelargonate ≥98%, halal

Synonyms: Ethyl nonanoate, Pelargonic acid ethyl ester, Nonanoic acid ethyl ester

W244708-1KG-K W244708-20KG-K W244708-9KG-K
220230-703EA 117.69 USD
220230-703 220230-704 220230-705
Ethyl pelargonate ≥98%, halal
Ethyl pelargonate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
MDL Number: MFCD00009570
CAS Number: 123-29-5
EINECS: 204-615-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR