Ethyl myristate ≥98% Kosher

Synonyms: Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester

W244503-1KG-K W244503-20KG-K W244503-9KG-K W244503-4KG-K
220230-698EA 241.54 USD
220230-698 220230-699 220230-701 220230-700
Ethyl myristate ≥98% Kosher
Ethyl myristate
Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00008984
CAS Number: 124-06-1
EINECS: 204-675-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR