Ethyl lactate (synthetic) ≥98% FCC, FG

Danger

Synonyms: 2-Hydroxypropionic Acid Ethyl Ester, Ethyl 2-Hydroxypropionate, Lactic Acid Ethyl Ester

W244007-1KG-K W244007-5KG-K W244007-25KG-K
220231-683EA 117.69 USD
220231-683 220231-685 220231-684
Ethyl lactate (synthetic) ≥98% FCC, FG
Ethyl lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154 °C (lit.)
Melting Pt: –25 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00065359
CAS Number: 97-64-3
EINECS: 202-598-0
UN: 1192
ADR: 3,III
Merck Index: 12,03850

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR