4-Ethyl-2-methoxyphenol ≥98% FCC, FG Kosher, Halal

Warning

Synonyms: 5-Ethyl-2-hydroxyanisole, 1-Hydroxy-2-methoxy-4-ethyl Benzene, 4-Ethylguaiacol

W243604-100G-K W243604-5KG-K W243604-1KG-K
220230-902EA 236.92 USD
220230-902 220230-904 220230-903
4-Ethyl-2-methoxyphenol ≥98% FCC, FG Kosher, Halal
4-Ethyl-2-methoxyphenol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00038714
CAS Number: 2785-89-9
EINECS: 220-500-4

Order Now

Specification Test Results

Appearance (Color) Colorless to Light Yellow
Appearance (Form) Liquid
Refractive index at 20 °C 1.525 - 1.530
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.00%
Specific Gravity 1.061 - 1.064
Solubility (Turbidity) Clear
Solubility (Color) 1ml/1ml,95% Ethanol Colorless to Faint Yellow
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Mercury (Hg) ≤1 ppm
Lead (Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR