Ethyl decanoate ≥98% FCC

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

W243213-1KG-K W243213-4KG-K W243213-100G-K
220231-531EA 286.15 USD
220231-531 220231-532 220231-530
Ethyl decanoate ≥98% FCC
Ethyl decanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: -20 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR