Ethyl cinnamate Kosher

Warning

Synonyms: NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester

W243000-1KG-K W243000-25KG-K W243000-10KG-K
220231-514EA 220 USD
220231-514 220231-515 220231-513
Ethyl cinnamate Kosher
Ethyl cinnamate
Formula: C₁₁H₁₂O₂
MW: 176.21 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.046 g/cm³ (20 °C)
MDL Number: MFCD00009189
CAS Number: 103-36-6
EINECS: 203-104-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR